Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

11H-Benzo[a]carbazole 98.0+%, TCI America™
SDP

Catalog No. B46351G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B46351G 1 g
B46355G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B46351G Supplier TCI America Supplier No. B46351G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 239-01-0
Molecular Formula C16H11N
Molecular Weight (g/mol) 217.271
MDL Number MFCD00215939
InChI Key MYKQKWIPLZEVOW-UHFFFAOYSA-N
Synonym 1,2-Benzocarbazole
PubChem CID 9196
IUPAC Name 11H-benzo[a]carbazole
SMILES C1=CC=C2C(=C1)C=CC3=C2NC4=CC=CC=C34

Specifications

Melting Point 233°C
Color White-Yellow
Quantity 1 g
Formula Weight 217.27
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 11H-Benzo[a]carbazole
RTECSNumber DE7025000
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.