Learn More
Apexbio Technology LLC 11Z,14Z-Eicosadienoic acid 5598-38-9 50mg
SDP
Supplier:  Apexbio Technology LLC A996650
Encompass_Preferred
11Z 14Z-Eicosadienoic acid (CAS 5598-38-9) is a polyunsaturated fatty acid of the -6 series with a 20-carbon chain It is naturally found in high concentrations in certain plant-derived oils such as safflower and sunflower oils Within biological systems this compound is not synthesized endogenously and must be obtained from the diet Functionally 11Z 14Z-Eicosadienoic acid serves as a metabolic precursor to arachidonic acid thereby influencing the biosynthesis of eicosanoids involved in inflammation immune regulation and signal transduction It is widely used in studies exploring lipid metabolism cell membrane dynamics and the role of dietary fatty acids in physiological and pathological processes
Catalog No. 50-000-96009
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.