Learn More
ProteinSimple 12-230 KDA WES SEPARATION MODU
Supplier:  ProteinSimple SMW003
Encompass
12-230 kDa Wes Separation Module, 2 x 25 capillary cartridges.Use with a Detection Module. Reagents for 50 data points includes two parts: Separation box with 2x Capillary Cartridges (25 capillaries per cartridge), 2x Wes 12-230 kDa pre-filled Plates (pre-filled with Separation Matrix 2, Stacking Matrix 2, Split Running Buffer 2 and Matrix Removal Buffer), Sample Buffer, Wash Buffer (Store at RT), plus 1x Standard Pack 1 clamshell with 2x DTT and 2x FL master mix with 29 kDa System Control protein. (Store at 4 C).
Catalog No. NC1461323