Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

12-Bromo-1-dodecanol 98.0+%, TCI America™
SDP

Catalog No. B173125G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B173125G 25 g
B17315G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B173125G Supplier TCI America Supplier No. B173125G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 3344-77-2
Molecular Formula C12H25BrO
Molecular Weight (g/mol) 265.24
MDL Number MFCD00004754
InChI Key ASIDMJNTHJYVQJ-UHFFFAOYSA-N
PubChem CID 137895
IUPAC Name 12-bromododecan-1-ol
SMILES OCCCCCCCCCCCCBr

Specifications

Melting Point 33°C
Color White-Yellow
Boiling Point 156°C
Quantity 25 g
Formula Weight 265.24
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 12-Bromo-1-dodecanol
TSCA No
Recommended Storage Refrigerator
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.