Learn More
National Safety Apparel™ Enespro™ ArcGuard™ 12 cal Arc Flash Kit with Balaclava and Voltage Gloves (09)
SDP
With a unique blend of 88% cotton and 12% high-tenacity nylon, garments provide a smooth feel for increased comfort and the inherently FR material delivers exceptional performance in reliability
$1409.00 - $1475.00
Specifications
Ankle Style Open
Arc Flash Rating 12 cal/cm²
Certifications/Compliance CAT 2, NFPA 70E, NFPA 2112, CSA Z462, ASTM F1506, OSHA 1910.269
Closure Type FR Hook and Loop
Color Navy
View More Specs
Products 8
Catalog Number Mfr. No. Size Price Quantity & Availability  
Catalog Number Mfr. No. Size Price Quantity & Availability  
17-001-1215
SDP
View Documents
National Safety Apparel™
KIT2SC11SM09
Small
Each for $1,409.00
Only null left
Add to Cart
17-001-1220
SDP
View Documents
National Safety Apparel™
KIT2SC11MD09
Medium
Each for $1,409.00
Only null left
Add to Cart
17-001-1225
SDP
View Documents
National Safety Apparel™
KIT2SC11LG09
Large
Each for $1,439.00
Only null left
Add to Cart
17-001-1230
SDP
View Documents
National Safety Apparel™
KIT2SC11XL09
X-Large
Each for $1,409.00
Only null left
Add to Cart
17-001-1235
SDP
View Documents
National Safety Apparel™
KIT2SC112X09
2X-Large
Each for $1,460.00
Only null left
Add to Cart
17-001-1240
SDP
View Documents
National Safety Apparel™
KIT2SC113X09
3X-Large
Each for $1,430.00
Only null left
Add to Cart
17-001-1245
SDP
View Documents
National Safety Apparel™
KIT2SC114X09
4X-Large
Each for $1,453.00
Only null left
Add to Cart
17-001-1250
SDP
View Documents
National Safety Apparel™
KIT2SC115X09
5X-Large
Each for $1,475.00
Only null left
Add to Cart
Description
Description
  • PureView™ faceshield technology features a greater range of visibility
  • Premium anti-fog and scratch resistant coating on faceshield
  • FR hook and loop front closure on short coat
  • Stand up collar for extra coverage
  • Rib knit cuffs for comfortable, secure fit
  • Quick disconnect and adjustable shoulder straps on bib overalls
  • Hook and loop at side of bib for easy access
  • Expandable leg opening to fit over boots
  • Made in the USA
Specifications
Specifications
Open
CAT 2, NFPA 70E, NFPA 2112, CSA Z462, ASTM F1506, OSHA 1910.269
Navy
Arc Flash Kit
None
12 cal/cm²
FR Hook and Loop
Cotton, Nylon
Stitched
Unisex
Videos
SDS
Documents
Documents
Product Certifications
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.