Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

National Safety Apparel™ Enespro™ 12 Cal ArcGuard™ Arc Flash Kit with Short Coat, Bib Overall with Lift Front Hood (12)
SDP

Catalog No. 170011263 Shop All National Public Seating Corporation Products
Change view
Click to view available options
Size:
Small
Medium
Large
X-Large
2X-Large
3X-Large
4X-Large
5X-Large
8 product options available for selection
Product selection table with 8 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
17-001-1263 Medium
17-001-1258 Small
17-001-1268 Large
17-001-1273 X-Large
17-001-1278 2X-Large
17-001-1283 3X-Large
17-001-1288 4X-Large
17-001-1293 5X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 8 options available.
8 options
Catalog No. 17-001-1263 Supplier National Safety Apparel™ Supplier No. KIT2SC11LFMD12
Add to Cart
Add to Cart

With a unique blend of 88% cotton and 12% high-tenacity nylon UltraSoft garments provide a smooth feel for increased comfort and the inherently FR material delivers exceptional performance in reliability

  • PureView™ faceshield technology features a greater range of visibility
  • Premium anti-fog and scratch resistant coating on faceshield
  • FR hook and loop front closure on short coat
  • Stand up collar for extra coverage
  • Rib knit cuffs for comfortable, secure fit
  • Quick disconnect and adjustable shoulder straps on bib overalls
  • Hook and loop at side of bib for easy access
  • Expandable leg opening to fit over boots
  • Made in the USA

Specifications

Certifications/Compliance CAT 2, NFPA 70E, NFPA 2112, CSA Z462, ASTM F1506, OSHA 1910.269
Gender Unisex
Material Cotton, Nylon
Product Type Arc Flash Kit
For Use With (Application) Utilities, Manufacturing, Electrical Contractors
Arc Flash Rating 12 cal/cm²
Closure Type FR Hook and Loop
Color Navy
Size Medium
Suspender None
Seam Type Stitched
Ankle Style Open
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.