Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

National Safety Apparel™ Enespro™ ArcGuard™ 12 cal Coverall Arc Flash Kit with Voltage Gloves (10)
SDP

Catalog No. 17001866 Shop All National Public Seating Corporation Products
Change view
Click to view available options
Size:
Small
Medium
Large
X-Large
2X-Large
3X-Large
4X-Large
5X-Large
8 product options available for selection
Product selection table with 8 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
17-001-866 X-Large
17-001-851 Small
17-001-856 Medium
17-001-861 Large
17-001-871 2X-Large
17-001-876 3X-Large
17-001-881 4X-Large
17-001-886 5X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 8 options available.
8 options
Catalog No. 17-001-866 Supplier National Safety Apparel™ Supplier No. KIT2CV11XL10
Add to Cart
Add to Cart

The premium choice for superior comfort, improved safety and above average durability

  • PureView™ faceshield technology features a greater range of visibility
  • Premium anti-fog and scratch resistant coating on faceshield
  • Action back allows for full range of motion
  • Flexible sewn-in elastic waist
  • 6 pocket style with side pass through access
  • Leg openings made to fit over work boots
  • Covered 2-way zipper

Specifications

For Use With (Application) Utilities, Manufacturing, Electrical Contractors
Certifications/Compliance CAT 2, ASTM F1506, OSHA 1910.269, NFPA 70E, NFPA 2112, CSA Z462
Gender Unisex
Product Type Arc Flash Kit
Arc Flash Rating 12 cal/cm²
Material Cotton, Nylon
Color Navy
Suspender None
Seam Type Stitched
Ankle Style Open
Size X-Large
Closure Type Zipper
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.