Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

National Safety Apparel™ Enespro™ ArcGuard™ 12 cal Coverall Arc Flash Kit with Lift Front Hood, No Gloves
SDP

Catalog No. 17001986 Shop All National Public Seating Corporation Products
Change view
Click to view available options
Size:
Small
Medium
Large
X-Large
2X-Large
3X-Large
4X-Large
5X-Large
8 product options available for selection
Product selection table with 8 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
17-001-986 Medium
17-001-985 Small
17-001-987 Large
17-001-988 X-Large
17-001-989 2X-Large
17-001-990 3X-Large
17-001-991 4X-Large
17-001-992 5X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 8 options available.
8 options
Catalog No. 17-001-986 Supplier National Safety Apparel™ Supplier No. KIT2CV11NGLFMD
Add to Cart
Add to Cart

The premium choice for superior comfort, improved safety and above average durability

  • FR Coverall
  • 11 in. Class 0 Rubber Voltage Insulating Gloves
  • 10 in. Leather Glove Protectors
  • Hard Hat with Arc Flash Faceshield and Chin Cup
  • Safety Glasses
  • Faceshield unit bag
  • Glove bag
  • Gear bag

Specifications

For Use With (Application) Utilities, Manufacturing, Electrical Contractors
Gender Unisex
Certifications/Compliance CAT 2, ASTM F1506, OSHA 1910.269, NFPA 70E, NFPA 2112, CSA Z462
Product Type Arc Flash Kit
Arc Flash Rating 12 cal/cm²
Material Cotton, Nylon
Color Navy
Size Medium
Suspender None
Ankle Style Open
Closure Type Zipper
Seam Type Stitched
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.