Learn More
National Safety Apparel™ Enespro™ ArcGuard™ 12 cal Coverall
Shop All National Public Seating Corporation Products

Description
- Comfortable sewn-in elastic waist for better fit
- Dual certified to protect against flash fire and arc flash hazards
- 2-way covered zipper front for easy entry
- Two front chest pockets with flaps and hidden snap closures
- Large reinforced back pockets
- Convenient 6 pocket style with side pass through access
- Leg openings made to fit over work boots
- All metal covered to ensure no contact with skin
- Action back allows for full range of motion
- FR fabric will not melt, drip, or ignite
- Zip front closure with additional covered snaps
Specifications
Specifications
| Product Type | Coveralls |
| Size | X-Large |
| Gender | Unisex |
| Material | Cotton, Nylon |
| Color | Navy |
| Disposable | Reusable |
| Waist Style | Elastic (Stitched-in) |
| Ankle Style | Open |
| Neck Style | Collared |
| Hood Type | Unhooded |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.