Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

National Safety Apparel™ Enespro™ ArcGuard™ 12 cal Coverall
SDP

Catalog No. 17001545 Shop All National Public Seating Corporation Products
Change view
Click to view available options
Size:
Small
Medium
Large
X-Large
2X-Large
3X-Large
4X-Large
5X-Large
8 product options available for selection
Product selection table with 8 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
17-001-545 2X-Large
17-001-541 Small
17-001-542 Medium
17-001-543 Large
17-001-544 X-Large
17-001-546 3X-Large
17-001-547 4X-Large
17-001-548 5X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 8 options available.
8 options
Catalog No. 17-001-545 Supplier National Safety Apparel™ Supplier No. C88UP2X32
Add to Cart
Add to Cart

Durable and comfortable ultrasoft coveralls offer a unique blend of 88% cotton and 12% high-tenacity nylon for a smooth feel. The inherently FR material delivers exceptional performance and reliability.

  • Comfortable sewn-in elastic waist for better fit
  • Dual certified to protect against flash fire and arc flash hazards
  • 2-way covered zipper front for easy entry
  • Two front chest pockets with flaps and hidden snap closures
  • Large reinforced back pockets
  • Convenient 6 pocket style with side pass through access
  • Leg openings made to fit over work boots
  • All metal covered to ensure no contact with skin
  • Action back allows for full range of motion
  • FR fabric will not melt, drip, or ignite
  • Zip front closure with additional covered snaps

Specifications

Product Type Coveralls
Size 2X-Large
Gender Unisex
Material Cotton, Nylon
Color Navy
Disposable Reusable
Waist Style Elastic (Stitched-in)
Ankle Style Open
Neck Style Collared
Hood Type Unhooded
Closure Type Zipper
Seam Type Stitched
Sterility Non-sterile
Protection Class CAT 2
Resistance Flame
Arc Flash Rating 12 cal/cm²
Certifications/Compliance ASTM F1506, OSHA 1910.269, NFPA 70E, NFPA 2112, CSA Z462
Suspender None
For Use With (Application) Utilities, Manufacturing, Electrical Contractors
Style Coveralls
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.