Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

United Scientific™ 12-Channel Micropipette
SDPSCI_ED

Catalog No. S39241 Shop All United Scientific Supplies, Inc Products
Change view
Click to view available options
Volume (Metric):
0.5 to 10 μL
10 to 100 μL
2 to 20 μL
20 to 200 μL
40 to 300 μL
5 to 50 μL
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Volume (Metric)
S39241 0.5 to 10 μL
S39242 2 to 20 μL
S39243 5 to 50 μL
S39244 10 to 100 μL
S39245 20 to 200 μL
S39246 40 to 300 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. S39241 Supplier United Scientific™ Supplier No. PMC1210
Only null left
Add to Cart
Add to Cart

The 12-Channel Micropipette is fully autoclavable and recommended for a wide variety of applications such as ELISA, molecular screening, kinetic studies, and DNA amplifications.

  • Multichannel micropipette.
  • Two-stop plunger.
  • Built-in tip ejector.
  • Autoclavable tip cone.
  • Click-stop adjustable digital display.
  • Easy to calibrate, maintain, and disassemble for autoclaving.
  • Manufactured in an ISO 9001:2008 certified facility.
  • Each unit calibrated according to ISO 8655 standards and includes a calibration certificate.

Specifications

Accuracy ±4.0% at 1.0 μL, ±2.5% at 5.0 μL, ±1.5% at 10.0 μL
Compatible Tips 200 μL
Adjustable Tip Spacing No
Autoclavable Autoclavable
Color White/Blue
Disposable Reusable
For Use With Micro Tip 200 μL
Grip Type Ergonomic
Material Plastic
Model PMC12-10
Number of Channels 12
Programmability No
Sterility Non-sterile
Type Micropipette
Volume (Metric) 0.5 to 10 μL
For Use With (Application) Electrophoresis, PCR
For Use With (Equipment) Micropipette Tips
Includes Twelve tips, one pipette holder
Increments 0.1 μL
Precision ≤4.0% at 1.0 μL, ≤2.5% at 5.0 μL, ≤1.5% at 10.0 μL
Warranty 1 Year
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.