Learn More
Research Products International Corp Twelve Channel Reagent Reservoir with Lid, Sterile, 10 per Package
SDP
Supplier:  Research Products International Corp 246275
Encompass
Ideal reagent reservoir for performing serial dilutions or pipetting different reagents with a multi-channel pipettor. Each clearly numbered chamber holds up to 3ml of reagent and has a ''V'' shaped bottom. Made from chemical resistant polypropylene that can also be autoclaved. Each reservoir is individually wrapped to insure cleanliness and supplied with a clear lid.
Catalog No. 50-550-239
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.