Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

National Safety Apparel™ Enespro™ High Voltage Leather Protectors
SDP

Catalog No. 17001635 Shop All National Public Seating Corporation Products
Change view
Click to view available options
Size:
8
9
10
11
12
Length (English):
10 in.
12 in.
10 product options available for selection
Product selection table with 10 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size Length (English)
17-001-635 10 10 in.
17-001-633 8 10 in.
17-001-668 8 12 in.
17-001-634 9 10 in.
17-001-669 9 12 in.
17-001-670 10 12 in.
17-001-636 11 10 in.
17-001-671 11 12 in.
17-001-637 12 10 in.
17-001-672 12 12 in.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 10 options available.
10 options
Catalog No. 17-001-635 Supplier National Safety Apparel™ Supplier No. DWH10L10
Add to Cart
Add to Cart

Glove protectors should be used with electrical insulating rubber gloves for additional protection against mechanical hazards.

  • Leather glove protectors should be used every time that a task requires electrical insulating gloves
  • 10 in. glove protectors are suitable for use with Class 0 to 00 electrical insulating rubber gloves
  • 12 in. glove protectors are suitable for use with Class 2 electrical insulating rubber gloves

Specifications

Certifications/Compliance ASTM F696, NFPA 70E, CSA Z462, OSHA 1910.269
Applications Industrial
Color White, Yellow
Hand Specific
Material Leather
Product Type Glove Protectors
Size 10
For Use With (Application) Utilities, Manufacturing, Electrical Contractors
For Use With (Equipment) Class 00 to 0 Electrical Insulating Rubber Gloves
Glove Cuff Style Elasticated
Series DWH10
Length (English) 10 in.
Length (Metric) 25.4 cm
Voltage 1000 V
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.