Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Estes™ Industries 12 in. Parachute
SDPSCI_ED

Catalog No. S13194 Shop All Estes Industries Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Length (English):
12 in.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Length (English)
S13194 12 in.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. S13194 Supplier Estes™ Industries Supplier No. 2264
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Preassembled parachute with printed two color design.

  • This 12 in. (30.5 cm) parachute is preassembled and ready to use
  • For use with model rockets that require a 12 in. parachute

Specifications

For Use With (Equipment) Model Rockets
Length (English) 12 in.
Size 12 in. (30.5 cm)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.