Learn More
Jackson Immuno Research Labs 12 NM COLLOIDAL GOLD-AFFINIPUR
Supplier: Jackson Immuno Research Labs 703205155
12 nm Colloidal Gold AffiniPure Donkey Anti-Chicken IgY (IgG) (H+L) (EM Grade)(min X Bov, Gt, GP, Sy Hms, Hrs, Hu, Ms, Rb, Rat, Shp Sr Prot)Target: ChickenHost: DonkeyAntibody Format: Whole IgGSpecificity: IgY (IgG) (H+L)Minimal Cross Reactivity: Bovine, Goat, Guinea Pig, Syrian Hamster, Horse, Human, Mouse, Rabbit, Rat, Sheep Serum ProteinsConjugate: 12 nm Colloidal GoldProduct Category: ImmunoGold ComplexesClonality: PolyclonalRRID: AB_2340368
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.