Learn More
Organomation 12 Position N-EVAP Nitrogen Evaporator
Supplier: Organomation 11155
The 12 position N-EVAP nitrogen evaporator (cat# 11155) is a compact unit that allows for up to 12 samples to be evaporated simultaneously. At each individual sample position, the gas flow can be controlled via a precision needle valve. This is ideal for the simultaneous evaporation of samples with varying evaporation end points. The sample tray has a spring tensioner which allows any test tube between 10-30 mm to be processed without any modifications to the instrument. An optional small sample holder allows samples with an outside diameter of 5-16 mm to be processed.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.