Learn More
Enzo Life Sciences 12(S)-HETE ELISA kit (5x96wells)
SDP
Supplier:  Enzo Life Sciences ADI901050
Encompass_Preferred
Sensitive ELISA kit enabling detection of 12(S)-HETE in culture supernatants and plasma from any species. The 12(S)-HETE EIA kit is a colorimetric competitive enzyme immunoassay kit with results in 5 hours. Absorbance is read at 405 nm. Fast, accurate, and reliable results are obtained with ready-to-use liquid color-coded reagents that reduce error. Alternative Name: 12(S)-hydroperoxy tetraenoic eicosatetraenoic acid. Sensitivity: 146 pg/ml (range 195 - 50,000 pg/ml). Assay Time: 5 hours. Applications: ELISA, Colorimetric detection. Species Reactivity: Species independent. Contents: GxR Microtiter plate, Conjugate, Antibody, Assay buffer concentrate, Wash buffer concentrate, Standard, pNpp Substrate, Stop solution
Catalog No. 50-658-214
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.