Learn More
Perkin Elmer US LLC 12 Volt Car Adapter for the Spectrum Two
Supplier: Perkin Elmer US LLC L9004128
The Car Adapter is a required component of the Portable Battery Pack for the Spectrum Two N (L1600233), but can be ordered separately as a replacement or spare using this part number.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.