Learn More
Cellvis 12 Well glass bottom plate with high performance #1.5 cover glass, 20/Pack
Supplier:  Cellvis P121.5HN
Encompass
12 well glass bottom plate with high performance #1.5 cover glass (0.170±0.005mm). Black frame, with lid. Individually packed. Designed for high resolution imaging such as confocal microscopy. Suitable for long term tissue culture. Manufactured in a class 100,000 clean room. Plate made from virgin polystyrene. German high quality cover glass of superior optical quality, glass thickness is 0.170±0.005mm. A USP class VI adhesive is used to assemble the cover glass and the plate. Sterilized by Gamma radiation.
Catalog No. NC0799106