Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich o-Vanillin

Catalog No. 120804500G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
12-080-4500G 500 g
12-080-4100G 100 g
12-080-410G 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 12-080-4500G Supplier Sigma Aldrich Supplier No. 120804500G
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

o-Vanillin

Specifications

CAS 148-53-8
Melting Point 40°C to 42°C (lit.)
Boiling Point 265°C to 266°C (lit.)
Molecular Formula C8H8O3
Linear Formula CH3OC6H3-2-(OH)CHO
MDL Number MFCD00003322
Quantity 500 g
Synonym 2-Hydroxy-3-methoxybenzaldehyde; 2-Hydroxy-m-anisaldehyde; 3-Methoxysalicylaldehyde
Molecular Weight (g/mol) 152.15
Percent Purity 99%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.