Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Socorex Qualitix™ 200 μL Micropipette Tips from DWK Life Sciences™
SDP

Catalog No. 12141512 Shop All DWK Life Sciences Products
Change view
Click to view available options
Format:
Bagged
Rack
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Format
12-141-512 Rack
12-141-511 Bagged
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 12-141-512 Supplier Socorex Supplier No. 3070.0200R
Add to Cart
Edge
Add to Cart

Certified free of nuclease, endotoxin and ATP

  • Wide selection, superior quality, autoclavable
  • Bags, racks and tipfill rack filling system
  • Lot by lot consistency
  • Reduced packaging size, less bench space requirement
  • Broad compatibility with major micropipette brands
TRUSTED_SUSTAINABILITY

Specifications

Volume (Metric) 200 μL
Filtered Non-filtered
Sterility Non-sterile
Color Natural
Autoclavable Autoclavable
Length (Metric) 49.6 mm
Certifications/Compliance Certified free of nuclease, endotoxin and ATP, RNase, DNase Free, Pyrogen Free
Clean Claims RNase, DNase, and pyrogen free
Format Rack
Tip Style Beveled
Packaging 10 x 96/Rack
Quantity 10 x 96/Rack (960 Tips)
Product Type Micropipette tips
For Use With (Equipment) Universal Fit Micro
Graduated Yes
Product Line Socorex Qualitix
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.