Learn More
Sino Biological Human S100A14 / S114 Protein (His Tag) 50ug
Supplier: Sino Biological 12251H07E50
A DNA sequence encoding the native human S100A14 (NP_065723.1) (Gly 2-His 104) was expressed, with a polyhistide tag at the N-terminus.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.