Learn More
Sino Biological Human PDK-1 Protein (His Tag) 50ug
Supplier:  Sino Biological 12312H07B50
Encompass_Preferred
A DNA sequence encoding the mature form of human PDK1 (NP_002601.1) (Ser 29-Ala 436) was expressed, with a polyhistidine tag at the N-terminus.
Catalog No. 50-161-7959
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.