Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Diethyl azelate

Catalog No. 124583100ML Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
12-458-3100ML 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 12-458-3100ML Supplier Sigma Aldrich Supplier No. 124583100ML
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Diethyl azelate

Specifications

CAS 624-17-9
Melting Point -16°C to +15.8°C
Density 0.973 g/mL (at 25°C)
Boiling Point 172°C (18 mmHg)
Molecular Formula C13H24O4
Refractive Index n20/D 1.435 (literature)
Linear Formula C2H5O2C(CH2)7 CO2C2H5
Quantity 100 mL
MDL Number MFCD00009218
Synonym Diethyl nonanedioate
Molecular Weight (g/mol) 244.33
Percent Purity 90%
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.