Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Axygen™ 1250μL Universal Pipette Tips
GREENER_CHOICE

Catalog No. 14222213 Shop All Axygen Products
Change view
Click to view available options
Packaging:
Bulk
Rack
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Packaging
14-222-213 Bulk
14-222-216 Rack
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 14-222-213 Supplier Axygen™ Supplier No. MTX1250C
Add to Cart
Add to Cart

Matrix compatible tips in clear color

  • Matrix-style tips fits Matrix Impact™ and Impact2™ brand pipettors
  • Maxymum Recovery™ tips allows complete retrieval of precious samples
  • Optimized tip bevel reduces sample hang-up and improves pipetting accuracy
  • Presterilized tips are exposed to e-beam radiation for complete security without degrading the plastic or performance
  • Volume range: 15 to 1250μL
  • Available in bulk or racked formats

TRUSTED_SUSTAINABILITY

Specifications

Volume (Metric) 1250 μL
Filtered Non-filtered
Sterility Non-sterile
Color Clear
Autoclavable Autoclavable
Model MTX-1250-C
Certifications/Compliance RNase, DNase-free
Clean Claims RNase/DNase-free, Nonpyrogenic, Human gDNA- and PCR-inhibition-free
Packaging Bulk
Quantity 500/Pk., 5000/Cs.
Product Type Pipette Tips
For Use With (Equipment) Thermo Scientific Matrix Impact
Graduated No
Material Polypropylene
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.