Learn More
Alkali Scientific 1250µl PurePoint™ Filter Pipet Tips, Sterile (8 racks of 96), 768/cs
SDP
Supplier:  Alkali Scientific FT1250
Encompass_Preferred
PurePoint™ Filter Pipet Tips are designed and manufactured using premium-grade 100% polypropylene for the most critical molecular biology liquid handling applications. All PurePoint™ Aerosol Filter Tips are gamma irradiated sterile and certified RNase/DNase-Free, human DNA-Free, and pyrogen-free. In addition, PurePoint™ Aerosol Filter Tips are Ultra-Clear with a Fine-Point design for the convenient, precise pipetting of precious nucleic acid and protein samples and are designed to be Universal-fit for the majority of research-grade pipettors*. *Not suitable for Rainin LTS (Light Touch) Pipettors.
Catalog No. NC1372388
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.