Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich 1,4-Cyclohexadiene

Catalog No. 12541525ML Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mL
25 mL
100 mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
12-541-525ML 25 mL
12-541-5100ML 100 mL
12-541-55ML 5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 12-541-525ML Supplier Sigma Aldrich Supplier No. 12541525ML
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

  • 1,4-cyclohexadiene is an effective hydrogen donor for catalytic hydrogenation reactions
  • It can rapidly replace benzyl groups of n-benzyloxycarbamates, benzyl esters, benzyl ethers and benzyl amines with hydrogen
  • It forms benzene at elevated temperatures in the presence of a ruthenium(ii)-triphenylphosphine catalyst.useful for the reduction of radical intermediates formed in electron-transfer mediated ring-opening reactions

Specifications

CAS 628-41-1
Density 0.847 g/mL (at 25°C (literature))
Boiling Point 88°C to 89°C (lit.)
Molecular Formula C6H8
Linear Formula C6H8
Refractive Index n20/D 1.472 (literature)
MDL Number MFCD00001535
Quantity 25 mL
Synonym 1,4-Dihydrobenzene
Molecular Weight (g/mol) 80.13
Percent Purity 97%
Biological Activity Blood
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.