Learn More
Description
Color coded for easy identification
The Selectfrost microscope slides include colored tabs that allow grouping according to urgency, test, hospital or physician.
- Autoclavable
- Available in an array of tab colors
- Durable, opaque tab is inert to common laboratory chemicals and reagents
- Does NOT include SPECIMEN orientation key printed below tab
- Packaged in paperboard box
- Size: 25 × 75 × 1mm
Specifications
Specifications
| Length (Metric) | 25 mm |
| Frosted | Selectfrost Color-coded |
| Width (Metric) | 75 mm |
| Type | Microscope Slide |
| Thickness (Metric) | 1 mm |
| Color | Blue |
| Corner Style | Plain |
| Quantity | 144/Pk. |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.