Learn More
Description
- 1,4-cyclohexanedimethano is extensively used as cross-linking reagent in polymer industry.1,4-cyclohexanedimethanol has been used in the synthesis of polyketal copolymers
- It was used as diol comonomer during the synthesis of polyester-carbonates based on 1,3-propylene-co-1,4-cyclohexanedimethylene succinate
Specifications
Specifications
| CAS | 105-08-8 |
| Boiling Point | 283°C (lit.) |
| Molecular Formula | C8H16O2 |
| Linear Formula | C6H10(CH2OH)2 |
| MDL Number | MFCD00001512 |
| Quantity | 3 kg |
| Synonym | 1,4-Bis(hydroxymethyl)cyclohexane, mixture of cis and trans |
| Molecular Weight (g/mol) | 144.21 |
| Percent Purity | 99% |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.