Learn More
Chemglass Life Sciences Flask, 125mL, Filtering, # 5 Rubber Stopper, With Hose Connection
SDP
Supplier:  Chemglass Life Sciences CG8522125
Encompass_Preferred
Flask, 125mL, Filtering, # 5 Rubber Stopper, With Hose Connection. These small filtering flasks are recommended for microchemical use. All flasks have permanent white enamel marking spots.PLEASE NOTE: FOR LARGER CAPACITY FILTERING FLASKS, SEE CG-8514
Catalog No. CG8522125
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.