Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich 2-Hydroxyethyl methacrylate

Catalog No. 12863518KG Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
500 g
1 kg
18 kg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
12-863-518KG 18 kg
12-863-5500G 500 g
12-863-51KG 1 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 12-863-518KG Supplier Sigma Aldrich Supplier No. 12863518KG
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

2-Hydroxyethyl methacrylate (HEMA) was the first monomer to be used to synthesize hydrogels for biomedical applications.The water swelling properties of HEMA are enhanced by copolymerization with more hydrophilic monomers.HEMA finds wide applications for drug delivery.HEMA is used in the synthesis of biologically functional poly (2-hydroxyethyl methacrylate) (PHEMA) coplymers. It is also used to prepare light responsive membranes of PHEMA, HEMA/ acrylamide based specific drug release hydrogel, and water soluble HEMA/methacrylic acid hydrogel for drug delivery. Effects of HEMA on the migration of dental pulp stem cells (in vitro) may be studied.

Specifications

CAS 868-77-9
Density 1.073 g/mL (at 25°C)
Boiling Point 67°C (3.5 mmHg)
Molecular Formula C6H10O3
Linear Formula CH2=C(CH3)COOCH2CH2OH
Refractive Index n20/D 1.453 (literature)
MDL Number MFCD00002863
Quantity 18 kg
Synonym 1,2-Ethanediol mono(2-methylpropenoate); Glycol methacrylate; HEMA
Molecular Weight (g/mol) 130.14
Percent Purity 97%
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.