Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MP Biomedicals™ 12mm Culture Tube Caps

Catalog No. MP07DP1050 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Material:
Polypropylene
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Material
MP07DP1050 Polypropylene
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MP07DP1050 Supplier MP Biomedicals™ Supplier No. 07DP1050
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Fits 12mm x 75mm culture tubes. MP Biomedicals 12mm Culture Tube Caps are snap-on, disposable lids made of robust polypropylene.

TRUSTED_SUSTAINABILITY

Specifications

Closure Material Polypropylene
Closure Size 12 mm
Closure Type Snap Cap
Material Polypropylene
For Use With (Equipment) 12 x 75 mm culture tubes
Sterility Non-sterile
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.