Learn More
Ace Glass, Inc. 12MM POR E TUBES 12MM POR E TUBES
SDP
Supplier:  Ace Glass, Inc. 720910
Encompass_Preferred
12mm filtering tube with porosity E (4-8 micron) fritted disc sealed on bottom. Overall length is 125mm.
Catalog No. 50-974-652
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.