Learn More
Thomas Scientific 0.2 mL 12-Strip PCR Tubes & Domed Strip Caps Packed Together, Assorted, 80/pk
SDP
Supplier:  Thomas Scientific 1149Z74
Encompass
Thin Wall PCR Strip Tubes & Caps - 12 Strip. Available with dome caps or optically clear flat caps (for real-time PCR). Available in clear, assorted, or white tubes (real-time compatible). Convenient combo pack of tubes and caps -- no need to purchase separately. Deep seating caps provide a reliable, consistent seal to eliminate evaporation. Thin walls allow for efficient heat transfer and superior yields. Certified RNase, DNase and Pyrogen free. Ultra-thin and uniform wall thickness allows for precise thermal transfer along the entire surface of the tube. Full-length, even insertion caps guarantee a perfect fit to ensure there is no sample evaporation during thermal cycling.
Catalog No. NC1672189