Learn More
Market Lab Instrument Vibration Mat 12" x 12" x 0.5"
Supplier:  Market Lab ML111398GY
Encompass_Preferred
The Instrument Vibration Mat by MarketLab is a 12" x 12" x 0.5" thick rubber mat designed to isolate delicate instruments from external vibrations. It features a ribbed bottom to prevent sliding and has a max load capacity of 75 lbs. The mat is made from 100% recycled rubber in an anti-static gray color. It can help reduce vibration transmission in tabletop experiments, microscopy, balances, and other sensitive instrumentation. The high quality rubber withstands temperatures from -40°F to 200°F.
Catalog No. NC2334963
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.