Learn More
Enzo Life Sciences 13,14-Dihydro-prostaglandin E1 (10mg). CAS: 19313-28-1
SDP
Supplier:  Enzo Life Sciences BMLPG0480010
Encompass_Preferred
Circulating metabolite of PGE1 in man. Vasodilator equipotent with PGE12. Inhibits human platelet aggregation. Antimitotic activity. Alternative name: 13,14-Dihydro-PGE1. Purity: ≥98% (TLC). Long Term Storage: -80°C.
Catalog No. 50-201-1423
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.