Learn More
Description
- 15-gauge aramid fiber shell
- Black textured nitrile palm
- Seamless knit
- Allows manual dexterity
- Durable and comfortable
Specifications
Specifications
| ANSI Abrasion Rating | ANSI 3 |
| Certifications/Compliance | ASTM F1060-18 |
| Construction | Seamless Knit |
| Color | Black, Yellow |
| Cut Resistance Rating | ANSI A2 |
| Material | Aramid Fiber |
| Product Type | Cut-resistant Gloves |
| Size | Large |
| For Use With (Application) | Abrasive applications, assembly, box handling, brick handling, construction, cut protection, farming, general maintenance, knife handling, landscaping, plumbing, recycling |
| Glove Cuff Style | Knit |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.