Learn More
Sigma Aldrich Fine Chemicals Biosciences 13-Propanesultone 98100G
Supplier:  Sigma Aldrich Fine Chemicals Biosciences P50706100G
Encompass
1 3-Propanesultone is a cyclic sulfonic ester mainly used to introduce a propane sulfonic functionality into the organic structure.
Catalog No. 50-289-7245
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.