Learn More
Brooks Automation 13 Way Error Rack
Catalog No. PSSRTP577 Shop All Brooks Automation Products
Change view
Click to view available options
Type:
Rack
Catalog No. Type
PSSRTP577 Rack
1 options
Catalog No. PSSRTP577 Supplier Brooks Automation Supplier No. PSSRTP577
Add to Cart
Add to Cart
Precision rack module for managing and isolating specimen tubes flagged with errors in PathFinder™ systems
  • The 13 Way Error Rack from Brooks Automation is a precision-designed accessory used in PathFinder™ laboratory automation systems to manage and isolate specimen tubes flagged with errors or exceptions during processing
  • Featuring 13 secure positions, this rack ensures proper organization and traceability of nonconforming samples, minimizing workflow disruptions and supporting quality control Built from durable, lab-grade materials, the rack integrates seamlessly with
  • Brooks PathFinder hardware to maintain efficiency, safety, and compliance in automated specimen handling environments
Specifications
Type Rack