Learn More
Cayman Chemical ArachIdonyl m-NItronIlIn 5mg
SDP
Supplier:  Cayman Chemical 900595 MG
Encompass_Preferred
An unsaturated ionizable cationic lipid has been used in the generation of LNPs for the delivery of mRNA in vivo intravenous administration of LNPs containing lipid 29d and encapsulating mRNA encoding a luciferase reporter induce bioluminescence in the lungs and spleen of mice
Catalog No. 50-289-5394
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.