Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich 1-Methylpiperazine

Catalog No. 130001500G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
13-000-1500G 500 g
13-000-1100G 100 g
13-000-15G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 13-000-1500G Supplier Sigma Aldrich Supplier No. 130001500G
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

  • 1-methylpiperazine was used as mimic template in the preparation of molecularly imprinted microspheres (mims)
  • It was also used to prepare the difunctional strong anion-exchange stationary phase from a 1,4-diazacyclohexane derivative

Specifications

CAS 109-01-3
Density 0.903 g/mL (at 25°C (literature))
Boiling Point 138°C (lit.)
Molecular Formula C5H12N2
Refractive Index n20/D 1.466 (literature)
Linear Formula C5H12N2
MDL Number MFCD00005966
Quantity 500 g
Molecular Weight (g/mol) 100.16
Percent Purity 99%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.