Learn More
Sino Biological Human IGSF11 / BTIGSF Protein (His Tag) 20ug
Supplier:  Sino Biological 13094H08H20
Encompass_Preferred
A DNA sequence encoding the extracellular domain of human IGSF11 (AAH34411.1) (Met 1-Gly 240) was fused with a polyhistidine tag at the C-terminus
Catalog No. 50-161-8253
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.