Learn More
Sino Biological Recombinant Aequorea victoria GFP Protein (His Tag), HPLC-verified
Supplier:  Sino Biological 13105S07E100
Encompass_Preferred
A DNA sequence encoding the Aequorea victoria GFP (AAB65663) (Ser2-Lys238) was expressed with a polyhistide tag at the N-terminus.
Catalog No. 50-161-8277