Learn More
Sino Biological Human ADK Protein (His & GST Tag) 50ug
Supplier:  Sino Biological 13149H20B50
Encompass_Preferred
A DNA sequence encoding the human ADK isoform short (AAH03568.1) (Met 1-His 345) was fused with the N-terminal polyhistidine-tagged GST tag at the N-terminus.
Catalog No. 50-161-8329
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.