Learn More
Description
- Trans-2-hexen-1-al was used in evaluation of quality of protected designation of origin virgin olive oils by headspace solid-phase microextraction-gas chromatography using flame ionization detection and multivariate analysis
- It was used in the multicomponent method for the determination of low-molecular-weight carbonyl compounds in biological samples
Specifications
Specifications
| CAS | 6728-26-3 |
| Density | 0.846 g/mL (at 25°C (literature)) |
| Boiling Point | 47°C (17 mmHg) |
| Molecular Formula | C6H10O |
| Refractive Index | n20/D 1.446 (literature) |
| Linear Formula | CH3CH2CH2CH=CHCHO |
| MDL Number | MFCD00007008 |
| Quantity | 25 g |
| Molecular Weight (g/mol) | 98.14 |
| Percent Purity | 98% |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.