Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich trans-2-Phenylcyclopropyl isocyanate

Catalog No. 1343175G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
250 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
13-431-75G 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 13-431-75G Supplier Sigma Aldrich Supplier No. 1343175G
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

trans-2-Phenylcyclopropyl isocyanate

Specifications

CAS 63009-74-5
Density 1.074 g/mL at 25°C (lit.)
Boiling Point 75°C to 76°°C (0.5 mmHg)
Molecular Formula C10H9NO
Refractive Index n20/D 1.546 (lit.)
Linear Formula C6H5C3H4NCO
MDL Number MFCD00001273
Quantity 250 mg
Molecular Weight (g/mol) 159.18
Percent Purity 90%
Biological Activity Respiratory System
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.