Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fisherbrand™ accumet™ Basic AB315 Benchtop Laboratory pH/mV Meter

Catalog No. 13636AB315A Shop All Fisherbrand Products
Change view
Click to view available options
Type:
pH/mV Meter
pH/mV Meter with Probe
pH/mV Meter with Probe, NIST
pH/mV Meter, NIST
Probe Type:
Gel-filled pH/ATC Electrode
Refillable pH Electrode
Material:
Epoxy Body
Glass Body
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Type Probe Type Material
13636AB315A pH/mV Meter with Probe Gel-filled pH/ATC Electrode Epoxy Body
13636AB315 pH/mV Meter - -
13636AB315B pH/mV Meter with Probe Refillable pH Electrode Glass Body
AB315CERT pH/mV Meter with Probe, NIST - -
AB315ACERT pH/mV Meter, NIST Gel-filled pH/ATC Electrode Epoxy Body
AB315BCERT pH/mV Meter with Probe, NIST Refillable pH Electrode Glass Body
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. 13636AB315A
Add to Cart
Add to Cart

Ideal for daily pH testing of your routine laboratory samples, the accumet™ Basic AB315 Benchtop Meter provides essential functions needed for common pH measurements without complicated functions, all at a budget-friendly price.

  • Large, backlit display simultaneous shows pH, mV and temperature readings
  • One to five point pH calibration with automatic buffer recognition of four pH buffer sets
  • One point relative mV calibration (RmV) for ORP measurements
  • One point ATC temperature probe calibration
  • Calibration due alarm, selectable 1 to 60 hr., or off
  • Automatic or manual temperature compensation
  • Ready icon indicates measurement stability
  • Electrode condition icon indicates electrode status 500 data set storage with time/date stamp
  • Non-volatile meter memory preserves settings and data in event of power loss
  • Meter-attached stand facilitates easy electrode movement into and out of samples
TRUSTED_SUSTAINABILITY

Specifications

Description pH/mV Meter with pH/ATC Epoxy-body Double Junction Gel-filled Electrode
Accuracy (Temperature) ±0.3°C (±0.5°F)
Accuracy (pH) ±0.01
Data Management Details Printed or CSV Format
Accuracy (mV) ±0.2 or ±0.05%, Whichever is Greater
Outputs Port for Computer or Printer
Includes pH/ATC Electrode, Electrode Stand, Power Adapter
Measurement Modes pH with mV and Temperature, Relative mV with mV and Temperature
Calibration pH, Relative mV, Temperature
Range (pH) -2 to 18
Resolution (pH) 0.01, 0.1
Calibration Points (pH) USA: 2, 4.01, 7, 10.01, 12; NIST: 1.68, 4.01, 6.87, 9.18, 12.46; DIN: 1.09, 3.06, 4.65, 6.79, 9.23, 12.75; FSCI: 1, 3, 6, 8, 10, 13
Range (mV) ±2000 mV
Resolution (mV) 1 mV, 0.1 RmV
Calibration Points 1 to 5 (pH), 1 (Relative mV), 1 (Temperature)
Temperature Range (Metric) -5°C to +105°C
Temperature Range (English) 23°F to 221°F
Resolution (Temperature) 0.1°C (0.1°F)
Display Type 5 in. LCD
Backlight Option Yes
Memory Non-volatile
Data Points 500
Log Function Type Automatic, Manual
Inputs BNC, ATC
Certifications/Compliance CE, TUV 3-1, FCC Class A, Without NIST-traceable Calibration Certificate
IP Rating IP54
AC Adapter Universal Power Adapter
Warranty 3 Years
Material Epoxy Body
Model AB315
Probe Type Gel-filled pH/ATC Electrode
Range (Relative mV) ±2000 mV
Stability Indicator Icons
Temperature Compensation Automatic, Manual
Channels 1
Type pH/mV Meter with Probe
Voltage 100/240 V
Frequency 50/60 Hz
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.