Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fisherbrand™ Paper pH Strips

Catalog No. 13640512 Shop All Fisherbrand Products
Change view
Click to view available options
pH Range:
0 to 14
0 to 3
10 to 12
12.5 to 14
3 to 5.5
6 to 8
8 to 9.5
Type:
pH Test Paper
pH Test Paper Set
8 product options available for selection
Product selection table with 8 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. pH Range Type
13-640-512 8 to 9.5 pH Test Paper
13-640-509 3 to 5.5 pH Test Paper
13-640-511 0 to 3 pH Test Paper
13-640-510 6 to 8 pH Test Paper
13-640-508 0 to 14 pH Test Paper
13-640-513 10 to 12 pH Test Paper
13-640-514 12.5 to 14 pH Test Paper
13-640-515 0 to 14 pH Test Paper Set
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 8 options available.
8 options
This item is not returnable. View return policy
Catalog No. 13-640-512
Add to Cart
Add to Cart
This item is not returnable. View return policy

An easy choice for classroom experiments or demonstrations

  • Ideal for educational use
  • Low-cost paper strips are easy to pass around the classroom
  • Easy-to-use—simply dip strip in solution to be tested and match to color chart immediately

Order Info

Box of 10 vials, 100 low-cost paper strips per vial.

Specifications

pH Range 8 to 9.5
For Use With (Application) For classroom experiments or demonstrations
Graduations 0.5
Quantity 100 Tests/Vial, 10 Vials/Cs.
Storage Requirements Room Temperature
Type pH Test Paper
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.