Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Orion™ All-in-One™ pH Buffer Kits

Catalog No. 13641420
Change view
Click to view available options
Description:
All-in-One pH Buffer Kit
ROSS All-in-One pH Buffer Kit
Standard All-in-One pH Buffer Kit
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Description
13-641-420 ROSS All-in-One pH Buffer Kit
09-357-72 All-in-One pH Buffer Kit
13-624-500 Standard All-in-One pH Buffer Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 13-641-420 Supplier Thermo Scientific™ Supplier No. 810199
Add to Cart
Add to Cart

Work with ease using this color-coded pH buffer kit that includes three buffers, storage solution and a storage bottle.

Thermo Scientific™ Orion™ All-in-One™ pH buffer kits include pH 4.01, 7.00 and 10.01 buffers that are color-coded for easy identification as well as storage solution to help ensure your pH electrode is properly maintained. Using high-quality pH buffers to calibrate your pH electrode and pH meter is the best way to ensure your pH measurements are accurate and reproducible. Orion pH buffers are NIST traceable and manufactured under ISO 9001 quality standards.

Kit includes one each:

  • 1 pint (475mL) bottle of pH 4.01 buffer (910104)
  • 1 pint (475mL) bottle of pH 7.00 buffer (910107)
  • 1 pint (475mL) bottle of pH 10.01 buffer (910110)
  • 1 pint (475mL) bottle of ROSS™ pH electrode storage solution (810001)
  • 1 oz. (60mL) bottle of general pH electrode cleaning solution (900024)
  • 12mm diameter pH electrode storage bottle


Click here for the most complete, up-to-date MSDS and certificates of analysis files for Orion solutions

TRUSTED_SUSTAINABILITY

Specifications

Description ROSS All-in-One pH Buffer Kit
For Use With (Application) Accurate calibrations and NIST traceability, laboratory and bench analysis
pH 4.01, 7.00, 10.01
Quantity 4 x 475 mL, 1 x 60 mL
Solution Type pH Buffer, Storage Solution and Cleaning Solution
Packaging Kit
Traceability to NIST Yes
Includes 475 mL bottle each of pH 4.01, 7.00 and 10.01 buffers; 475 mL bottle of ROSS pH electrode storage solution (810001); 60 mL bottle of pH electrode cleaning solution; and 12 mm diameter electrode storage bottle

Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.