Learn More
Sino Biological Recombinant Human STC2 Protein (His Tag)
Supplier:  Sino Biological 13653H08H100
Encompass_Preferred
A DNA sequence encoding the human STC2 (O76061) (Met1-Arg302) was expressed with a C-terminal polyhistidine tag.
Catalog No. 50-161-8743